Type:
Whole Allergen
Whole Allergen
Whole Allergen
Horse meat
f321
Equus spp.
XE5VC
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Horse meat is often included in meat products and may also appear on restaurant menues as beef.
Undeclared ingredient in meat products.
Sometimes served as beef in restaurants.
In a study of beef skin test positive atopic children six out of 12 children were also positive to horse meat.
Horse meat was commonly eaten during the early history of the domesticated horse but its use decreased during the Roman era and has almost ceased during Christian time. Today horse meat is eaten mainly in France, Italy, Spain and on Iceland. However, horse meat is often included in meat products and may also appear on restaurant menues as beef. Six out of 12 beef skin test positive atopic children where also positive to horse meat (1).
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.