Type:
Whole Allergen
Whole Allergen
Whole Allergen
Sweet gum
t211
Altingiaceae
Liquidambar styraciflua
Sweet gum tree, American sweetgum, Redgum
XM1FG7
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Sweet gum is native to North America and to scattered locations in northeastern and central Mexico, Guatemala, Belize, El Salvador, Honduras, and Nicaragua. Sweet gum has been introduced into California as well as southwestern British Columbia (1). It is now found planted all over the world.
Medicinally, Sweet gum is known as "Copalm balsam"; the resinous gum is used extensively in Mexico and Europe as a substitute for storax. Ointments and syrups are made from it. It is also used as a perfuming agent in soap
Asthma has not been reported (1).
In a study in Westchester County in the state of New York, out of 100 patients referred for allergic rhinitis, 65% had a positive skin prick test to at least 1 aeroallergen out of 48 in the test panel. Sweet gum was positive in 16 (2)
No allergens from this plant have yet been characterized.
An extensive cross-reactivity among the different individual species of the genus could be expected (3).
Last Reviewed- April 2022