Whole Allergen
t211 Sweet gum
t211 Sweet gum Scientific Information
Type:
Whole Allergen
Display Name:
Sweet gum
Allergen code:
t211
Family:
Altingiaceae
Latin Name:
Liquidambar styraciflua
Other Names:
Sweet gum tree, American sweetgum, Redgum
WHO/ICD-11 code:
XM1FG7
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Environmental Characteristics
Worldwide distribution 
Sweet gum is native to North America and to scattered locations in northeastern and central Mexico, Guatemala, Belize, El Salvador, Honduras, and Nicaragua. Sweet gum has been introduced into California as well as southwestern British Columbia (1). It is now found planted all over the world.
Medicinally, Sweet gum is known as "Copalm balsam"; the resinous gum is used extensively in Mexico and Europe as a substitute for storax. Ointments and syrups are made from it. It is also used as a perfuming agent in soap
Clinical Relevance
Asthma has not been reported (1).
In a study in Westchester County in the state of New York, out of 100 patients referred for allergic rhinitis, 65% had a positive skin prick test to at least 1 aeroallergen out of 48 in the test panel. Sweet gum was positive in 16 (2)
Molecular Aspects
No allergens from this plant have yet been characterized.
Cross-reactivity
An extensive cross-reactivity among the different individual species of the genus could be expected (3).
Compiled By
Last Reviewed-  April 2022
References
  1. Weber RW. Liquidambar styraciflua. Ann Allergy Asthma Immunol. 2003;90(6):A6.
  2. Basak P, Arayata R, Brensilver J Prevalence of specific aeroallergen sensitivity on skin prick test in patients with allergic rhinitis in Westchester County. Internet J Asthma Allergy Immunol 2008; 6(2)
  3. L. Y. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. Pharmacia Diagnostics AB Uppsala Sweden. 1982.