Paul, a 14-year-old boy with a history of severe rhino-conjunctivitis, visits his healthcare provider after two episodes of asthma during late spring and summer. Paul also experiences oral allergy syndrome when eating various fruits and vegetables (tomatoes, melon, mandarin, kiwi, and peaches). His parents want Paul to undergo specific immunotherapy (SIT).
Paul's healthcare provider conducts a full clinical history and physical examination and decides to test using grass pollen components.
Paul has a history of rhino-conjunctivitis, episodes of asthma, and oral allergy syndrome (OAS) when eating some fruits and vegetables.
Test |
Type |
Paul's Results |
Egg |
Whole Allergen |
0 |
Milk |
Whole Allergen |
0 |
Grass Mix |
Whole Allergen |
+3 |
Birch |
Whole Allergen |
+2 |
Paul’s previous healthcare provider recommended that he use oral antihistamines and nasal steroids during pollen season.
These results together with this patient's case history and symptoms, help confirm the diagnosis.
| Allergen | Type |
Paul's Results |
Timothy |
Whole Allergen |
11.1 |
Birch |
Whole Allergen |
8.3 |
Tomato |
Whole Allergen |
4.2 |
Melon |
Whole Allergen |
7.1 |
Peach |
Whole Allergen |
3.9 |
Kiwi |
Whole Allergen |
6.5 |
| Allergen | Type |
Paul's Results |
Phl p 1 |
Allergen Component |
10.4 |
Phl p 5b |
Allergen Component |
3.9 |
Phl p 6 |
Allergen Component |
1.1 |
Phl p 12 (profilin) |
Allergen Component |
8.7 |
Bet v 2 (profilin) |
Allergen Component |
6.3 |
The significant IgE levels to the grass-specific components Phl p 1 and 5b indicate a “true” grass pollen sensitization. Paul’s IgE response to profilin also supports the noted OAS to various fruits and vegetables.1
REFINED DIAGNOSIS
HEALTHCARE PROVIDER MANAGEMENT PLAN
Paul's healthcare provider advises him to:
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.