Mucor racemosus Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
While initially white or yellow in color with a fluffy texture, Mucor mold colonies grow rapidly and turn a dark or olive-gray color as spores develop.1 If allergic individuals inhale these spores, they may experience allergy symptoms.2 Mucor are often found in soil, plant debris, and dung.3 However, the mold also grows in indoor environments, as colonies have been found in homes, schools, and offices in mediums such as house dust, mattresses, carpet, and ventilation ducts.1
Mucor can cause a wide range of human diseases, known as mucormycoses. While the diseases are often fatal, they usually only occur in people already suffering from a separate disease.1 Worldwide, Mucor prevalence ranges widely. For example, 40 percent of allergic Scandinavian patients showed Mucor sensitivity while only 3.8 percent of asthmatics in the Netherlands experienced Mucor sensitivity during skin testing.4
Mucor is often found in soil, plant debris, stored grains, dairy products, and dung, as well as on plants and decaying fruits and vegetables.1,3 However, mold reproduces via spores, which can be transported by air, water, and insects.5,10 So even if a fungus originates outdoors, it often can enter a dwelling through a variety of means, including doorways, windows, vents, and heating and air conditioning systems.11 Mucor colonies have been found in homes, hospitals, schools, and offices in mediums such as house dust, carpet, mattresses, and ventilation ducts.1,12 Although some species can grow in temperatures up to 42 C (roughly 108 F), which means they may live within animals and humans, Mucor racemosus doesn't thrive in temps higher than 32 C (roughly 89 F).1,4 So although this species can cause infections, they're usually limited to the skin, which is cooler than the body's interior.1 Plus, Mucor is used in the production of fermented foods and beverages in Asia.4
Some people with Mucor racemosus allergy may also experience symptoms when exposed to other types of mold. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.13
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
If you are allergic to mold, your healthcare provider may recommend a plan that includes the following.2,5,15,16
Your healthcare provider may direct you to take one of the following medications to improve your allergy symptoms:
Mold allergy symptoms can range from mild to severe and vary person to person.2 Reactions can happen almost immediately after exposure, or they can be delayed. Symptoms are most common in mid-summer to early fall, but since molds grow both indoors and out, allergic reactions can occur all year.5
Symptoms typically include one or more of the following:2,6
Most mold allergy reactions involve the preceding symptoms.2 However, mold sensitization is also a major risk factor for developing upper and lower respiratory diseases such as asthma.7
A small percentage of asthmatics with inhaled mold allergy can also develop allergic urticaria (aka hives) when they eat or drink anything containing yeast or mold.8 In addition to causing allergic reactions, molds can also lead to infections along with toxic reactions.9
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
There are no credible reports in medical literature documenting indoor exposure to molds as a cause of anaphylaxis.14 However, exposure and sensitization to fungal allergens can promote the development and worsening of allergic diseases such as allergic rhinitis and asthma.13