Scores table
-
Algorithm scores are displayed for each sample across the four gene sets assayed. Values indicate gene expression levels for each set of cell type-specific markers, relative to the reference standards.
-
(Optional) Click Download as JPG to save a .jpeg file of the output for your records or for submission as a manuscript figure.