Scores box plots
Select from 1–12 samples to be plotted at one time.
-
Hover over the box plot to display the algorithm scores for the samples and reference values.
-
The box represents the range of values from the median value of the upper 50% of data points (Q1) to the median value of the bottom 50% of data points (Q3). Thus the box represents the range of values for the 50% of data points closest to the overall median value (Median:) of the sample data set. The horizontal line within the box represents the median value for the entire sample data set.
-
The whiskers represent the calculated maximum (Max:) and minimum (Min:) values of the data set.