We provide optimized, streamlined kits to help you prepare samples for Sanger sequencing and fragment analysis applications on Applied Biosystems™ Genetic Analyzers, as well as a comprehensive selection of standards, capillaries, polymers, and other accessories for your genetic analyzer.
Use the tabs below to find the kits, reagents, and accessories to support your applications.
New–ExoSAP-IT Express reagent: PCR cleanup in 5 minutes
We continue to develop superior Sanger sequencing kits that help provide solutions for all your applications and templates. Click on the links below to order Applied Biosystems™ sequencing kits.
Learn more about our sequencing kits
Finding sequencing primers is simple and fast with our new Invitrogen™ Primer Designer™ tool for Sanger sequencing, containing >300,000 predesigned primer pairs for PCR and sequencing.
View predesigned primers for Sanger sequencing
To design your own custom primers for PCR and Sanger sequencing, use the Oligo Perfect Designer.
PCR and Sequencing Primers
Order PCR and sequencing primers
The Applied Biosystems™ CleanSweep™ PCR Purification Reagent is used for enzymatic clean-up of an amplified PCR product in an easy-to-use, single pipetting step format. The reagent dephosphorylates unincorporated nucleotides and digests unused primers to ensure accurate downstream analysis of sequencing or single-nucleotide polymorphism (SNP) data.
Order CleanSweep PCR Purification reagent
Which sequencing reaction cleanup product is right for you?
Cleanup Method Description & Application Product
Reagent-Based Sequencing Template Cleanup Scavenges all unincorporated Applied Biosystems™ BigDye™ terminators. Stabilizes samples before analysis. Best for long and short fragment recovery.
BigDye XTerminator Purification Kit
Gel Filtration Spin Columns Recovers DNA fragments larger than 16 base pairs. Removes > 98% of salts, NTP's and other unwanted low-molecular-weight impurities.
Centri-Sep 96-Well Plates
Centri-Sep 8-Well Strips
Centri-Sep Columns
Learn more about sequencing reaction clean up ›
Fragment analysis techniques allow the development of multiple applications on the Genetic Analyzer. For an overview of the different applications visit the Fragment Analysis page.
We offer several kits for specific Applied Biosystems™ fragment analysis–based applications.
Fragment Analysis Kits
Kits and Reagents Applications Associated Reagents
SNaPshot Multiplex System Genotyping, BAC fingerprint, Methylation analysis SNaPshot primer focus kits, GeneScan 120 LIZ Size Standard
AFLP Analysis System Genome fingerprinting for microorganism or
Genome fingerprinting for Plant
Custom labeled primer Microsatellite, LOH, chimerism, ISSR analysis, SSCP, RFLP, TRFLP, MSMSA  
Fragment Analysis Size Standards
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ™
SNaPshot™
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan 500 ROX Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC™, NED (Dye Set DS-31 - Filter D**) ROX GeneScan 500 ROX Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM, JOE, NED (Dye Set DS-32 - Filter F) ROX GeneScan 500 ROX Size Standard
Microsatellite (Forensic)
AFLP
6-FAM, VIC, NED, PET™ (Dye Set DS-33 - Filter G5) LIZ GeneScan 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Pathogen Detection
Custom Fragment Analysis
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.