Product Image
Applied Biosystems™
GeneScan™ 500 ROX™ dye Size Standard
The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43103619,600 injections
401734800 Reactions
Catalog number 4310361
Price (BRL)
57.063,46
Each
Quantity:
9,600 injections
Price (BRL)
57.063,46
Each
The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments by electrophoresis. GeneScan™ 500 ROX™ Size Standard is designed for sizing DNA fragments in the 35-500 bp range and provides 16 single-stranded ROX™ dye labeled fragments of 35, 50, 75, 100, 139, 150, 160, 200, 250, 300, 340, 350, 400, 450, 490, and 500 bases. Each of the DNA fragments is labeled with a proprietary fluorophore, which results in a single peak when run under both denaturing and non-denaturing conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionGeneScan™ 500 ROX™ dye Size Standard, ROX™ Label. Use with 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
For Use With (Application)Sequencing
For Use With (Equipment)3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Label or DyeROX
Product TypeDye
Quantity9,600 injections
Run TimeStandard
Product LineGeneScan, ROX
Unit SizeEach
Contents & Storage
This kit contains ROX size standard and loading buffer.
Store the kit at 2°C to 8°C. (Do not freeze).