Search
Search

| Assay Name: | bta-miR-93 |
|---|---|
| miRBase Accession Number: | MI0005050 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAAGUGCUGUUCGUGCAGGUA |
| Species: | Bovine |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUGUUCGUGCAGGUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-93 | MIMAT0003837 |