Search
Search

| Assay Name: | hsa-miR-483-5p |
|---|---|
| miRBase Accession Number: | MI0002467 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGACGGGAGGAAAGAAGGGAG |
| Species: | Human, Gorilla gorilla |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | AAGACGGGAGGAAAGAAGGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-483-5p | MIMAT0004761 |
| Gorilla gorilla | ggo-miR-483 | MIMAT0024191 |