Search
Search

| Assay Name: | hsa-miR-15b-3p |
|---|---|
| miRBase Accession Number: | MI0000438 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGAAUCAUUAUUUGCUGCUCUA |
| Species: | Human, Mouse, Rat, Dasypus novemcinctus, Pteropus alecto, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | CGAAUCAUUAUUUGCUGCUCUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-15b-3p | MIMAT0004586 |
| Mouse | mmu-miR-15b-3p | MIMAT0004521 |
| Rat | rno-miR-15b-3p | MIMAT0017093 |
| Dasypus novemcinctus | dno-miR-15b-3p | MIMAT0047504 |
| Pteropus alecto | pal-miR-15b-3p | MIMAT0039906 |
| Rhesus monkey | mml-miR-15b-3p | MIMAT0026562 |