Search
Search

| Assay Name: | oan-miR-383-5p |
|---|---|
| miRBase Accession Number: | MI0006900 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGAUCAGAAGGUGAUUGUGGC |
| Species: | Ornithorhynchus anatinus, Sarcophilus harrisii |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000137, mir-383 |
Mature miRNA Sequence: | CAGAUCAGAAGGUGAUUGUGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-383-5p | MIMAT0007189 |
| Sarcophilus harrisii | sha-miR-383 | MIMAT0022829 |