Search
Search

| Assay Name: | cpo-miR-361-3p |
|---|---|
| miRBase Accession Number: | MI0038742 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCCCCAGGUGUGAUUCUGAUUUGC |
| Species: | Dasypus novemcinctus, Guinea pig, Papio hamadryas, Pig, Rabbit |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CCCCCAGGUGUGAUUCUGAUUUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dasypus novemcinctus | dno-miR-361-3p | MIMAT0047830 |
| Guinea pig | cpo-miR-361-3p | MIMAT0047189 |
| Papio hamadryas | pha-miR-361 | MIMAT0049511 |
| Pig | ssc-miR-361-3p | MIMAT0013934 |
| Rabbit | ocu-miR-361-3p | MIMAT0048394 |