Search
Search

| Assay Name: | dme-miR-9b-3p |
|---|---|
| miRBase Accession Number: | MI0000415 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGAGCUUUAUUACCAAAAACC |
| Species: | Drosophila melanogaster, Drosophila virilis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000014, mir-9 |
Mature miRNA Sequence: | UAGAGCUUUAUUACCAAAAACC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-9b-3p | MIMAT0020829 |
| Drosophila virilis | dvi-miR-9b-3p | MIMAT0032055 |