Search
Search

| Assay Name: | cel-miR-266 |
|---|---|
| miRBase Accession Number: | MI0000346 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCAAGACUUUGGCAAAGC |
| Species: | Caenorhabditis elegans |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000281, mir-266 |
Mature miRNA Sequence: | AGGCAAGACUUUGGCAAAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-266 | MIMAT0000322 |