Search
Search

| Assay Name: | mmu-miR-10b-3p |
|---|---|
| miRBase Accession Number: | MI0000221 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGAUUCGAUUCUAGGGGAAUA |
| Species: | Mouse, Ophiophagus hannah |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | CAGAUUCGAUUCUAGGGGAAUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-10b-3p | MIMAT0004538 |
| Ophiophagus hannah | oha-miR-10c-3p | MIMAT0036665 |