Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12718
          Assay Name: hsa-miR-202-3p
          miRBase Accession Number: MI0003130
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 133247511 - 133247620 [-] on Build GRCh38
          Mature miRNA Sequence: AGAGGUAUAGGGCAUGGGAA
          Species: Human, Pan troglodytes, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12718

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000121, mir-202

          Mature miRNA Sequence:

          AGAGGUAUAGGGCAUGGGAA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-202-3p MIMAT0002811
          Pan troglodytes ptr-miR-202 MIMAT0008064
          Taeniopygia guttata tgu-miR-202 MIMAT0014565

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-202
          Stem-loop Accession # MI0003130
          Stem-loop Sequence
          CGCCUCAGAGCCGCCCGCCGUUCCUUUUUCCUAUGCAUAUACUUCUUUGAGGAUCUGGCCUAAAGAGGUAUAGGGCAUGGGAAAACGGGGCGGUCGGGUCCUCCCCAGCG
          Chromosome Location Chr. 10 - 133247511 - 133247620 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002811
          miRBase ID: hsa-miR-202-3p
          Mature miRNA Sequence: AGAGGUAUAGGGCAUGGGAA
          Chromosome Location: Chr. 10 - 133247511 - 133247620 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-202
          Stem-loop Accession # MI0008574
          Stem-loop Sequence
          CUCCUCAGAGCCGCCCGCCGUUCCUUUUUCCUAUGCAUAUACUUCUUUGAGGAUCUGGCCUAAAGAGGUAUAGGGCAUGGGAAAACGGGGCGGUCGGGUCCUCCCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008064
          miRBase ID: ptr-miR-202
          Mature miRNA Sequence: AGAGGUAUAGGGCAUGGGAA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-202-1
          Stem-loop Accession # MI0013785
          Stem-loop Sequence
          CUUUUUCCUAUGCAUAUACUUCUUUGAGAAUUGCUUCUAAAGAGGUAUAGGGCAUGGGAAAAUGGGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014565
          miRBase ID: tgu-miR-202
          Mature miRNA Sequence: AGAGGUAUAGGGCAUGGGAA
          Chromosome Location: NA

          Stem-loop ID tgu-mir-202-2
          Stem-loop Accession # MI0013786
          Stem-loop Sequence
          CUUUUUCCUAUGCAUAUACUUCUUUGAGAAUUGCUUCUAAAGAGGUAUAGGGCAUGGGAAAAUGGGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014565
          miRBase ID: tgu-miR-202
          Mature miRNA Sequence: AGAGGUAUAGGGCAUGGGAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303863_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12718
          Ambion® Pre-miR™ miRNA Precursor : PM12718
          TaqMan™ MicroRNA Assay : 002363
          mirVana® miRNA mimic : MC12718
          TaqMan™ Advanced miRNA Assay : 478417_mir


          miRBase Alias:

          Human: hsa-miR-202 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-g6tbm:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0