Search
Search

| Assay Name: | hsa-miR-491-3p |
|---|---|
| miRBase Accession Number: | MI0003126 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUAUGCAAGAUUCCCUUCUAC |
| Species: | Human, Mouse, Dasypus novemcinctus, Goat, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000319, mir-491 |
Mature miRNA Sequence: | CUUAUGCAAGAUUCCCUUCUAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-491-3p | MIMAT0004765 |
| Mouse | mmu-miR-491-3p | MIMAT0017255 |
| Dasypus novemcinctus | dno-miR-491-3p | MIMAT0047925 |
| Goat | chi-miR-491-3p | MIMAT0036252 |
| Horse | eca-miR-491-3p | MIMAT0013117 |
| Pan troglodytes | ptr-miR-491 | MIMAT0008166 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-491-3p | MIMAT0015912 |
| Rhesus monkey | mml-miR-491-3p | MIMAT0006353 |