Search
Search

| Assay Name: | odi-miR-1481 |
|---|---|
| miRBase Accession Number: | MI0007092 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCACAAUGAAGAUAGAAGAUGGCU |
| Species: | Oikopleura dioica |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | CCACAAUGAAGAUAGAAGAUGGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Oikopleura dioica | odi-miR-1481 | MIMAT0006033 |