Search
Search

| Assay Name: | dme-miR-968-5p |
|---|---|
| miRBase Accession Number: | MI0005825 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAGUAGUAUCCAUUAAAGGGUUG |
| Species: | Drosophila melanogaster, Drosophila simulans |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000865, mir-968 |
Mature miRNA Sequence: | UAAGUAGUAUCCAUUAAAGGGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-968-5p | MIMAT0005484 |
| Drosophila simulans | dsi-miR-968-5p | MIMAT0012441 |