Search
Search

| Assay Name: | csa-miR-155 |
|---|---|
| miRBase Accession Number: | MI0007198 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAAUGCUAAUAAGUGAUUUAUG |
| Species: | Ciona savignyi |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UUAAUGCUAAUAAGUGAUUUAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ciona savignyi | csa-miR-155 | MIMAT0006131 |