Search
Search

| Assay Name: | isc-miR-285 |
|---|---|
| miRBase Accession Number: | MI0012275 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUUUGAAUUCAGUUC |
| Species: | Ixodes scapularis, Parasteatoda tepidariorum |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUUUGAAUUCAGUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ixodes scapularis | isc-miR-285 | MIMAT0012695 |
| Parasteatoda tepidariorum | pte-miR-29a-3p | MIMAT0046133 |