Search
Search

| Assay Name: | sja-miR-190-3p |
|---|---|
| miRBase Accession Number: | MI0015294 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUGACCAGACAUAUCCCU |
| Species: | Schistosoma japonicum, Schistosoma mansoni |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001853, mir-190_2 |
Mature miRNA Sequence: | CAGUGACCAGACAUAUCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schistosoma japonicum | sja-miR-190-3p | MIMAT0016266 |
| Schistosoma mansoni | sma-miR-190-3p | MIMAT0025028 |