Search
Search

| Assay Name: | api-miR-3055 |
|---|---|
| miRBase Accession Number: | MI0014018 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCAGUUCGCGAUAAGCGGCCG |
| Species: | Acyrthosiphon pisum |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001909, mir-3055 |
Mature miRNA Sequence: | GCAGUUCGCGAUAAGCGGCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Acyrthosiphon pisum | api-miR-3055 | MIMAT0014798 |