Search
Search

| Assay Name: | ath-miR162a-5p |
|---|---|
| miRBase Accession Number: | MI0000194 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAGGCAGCGGUUCAUCGAUC |
| Species: | Arabidopsis, Citrus sinensis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000127, MIR162_1 |
Mature miRNA Sequence: | UGGAGGCAGCGGUUCAUCGAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR162a-5p | MIMAT0031876 |
| Arabidopsis | ath-miR162b-5p | MIMAT0031877 |
| Citrus sinensis | csi-miR162-5p | MIMAT0017388 |