Search
Search

| Assay Name: | ath-miR158a-5p |
|---|---|
| miRBase Accession Number: | MI0000188 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUUGUCUACAAUUUUGGAAA |
| Species: | Arabidopsis, Arabidopsis lyrata |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000294, MIR158 |
Mature miRNA Sequence: | CUUUGUCUACAAUUUUGGAAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR158a-5p | MIMAT0031873 |
| Arabidopsis lyrata | aly-miR158a-5p | MIMAT0017421 |