Search
Search

| Assay Name: | dme-miR-992-5p |
|---|---|
| miRBase Accession Number: | MI0005852 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUAUCAGCAAAGUGUUAUUU |
| Species: | Drosophila melanogaster |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000914, mir-992 |
Mature miRNA Sequence: | UGGUAUCAGCAAAGUGUUAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-992-5p | MIMAT0020891 |