Search
Search

| Assay Name: | aca-miR-153-3p |
|---|---|
| miRBase Accession Number: | MI0018762 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGAU |
| Species: | Anolis carolinensis, Gadus morhua |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-153-3p | MIMAT0021786 |
| Gadus morhua | gmo-miR-153a-3p | MIMAT0044210 |