Search
Search

| Assay Name: | bdi-miR7747-5p |
|---|---|
| miRBase Accession Number: | MI0025365 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUCAGAUUGUUUUGUUGUAGGAUG |
| Species: | Brachypodium distachyon |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AUCAGAUUGUUUUGUUGUAGGAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Brachypodium distachyon | bdi-miR7747-5p | MIMAT0030234 |