Search
Search

| Assay Name: | pal-miR-361-3p |
|---|---|
| miRBase Accession Number: | MI0032462 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCCCAGGUGCGAUUCUGAU |
| Species: | Pteropus alecto |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCCCCCAGGUGCGAUUCUGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pteropus alecto | pal-miR-361-3p | MIMAT0040040 |