Search
Search

| Assay Name: | dno-miR-660-3p |
|---|---|
| miRBase Accession Number: | MI0039164 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCUCCUGUGUGCAUGGAUUACA |
| Species: | Dasypus novemcinctus |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | ACCUCCUGUGUGCAUGGAUUACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dasypus novemcinctus | dno-miR-660-3p | MIMAT0047995 |