Search
Search

Camk2a| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286809.1 | NP_001273738.1 | 1 | 392 | ||
| NM_009792.3 | NP_033922.1 | 1 | 392 | |||
| NM_177407.4 | NP_803126.1 | 13 | 1113 | |||
| GenBank mRNA | AK083245.1 | - | 13 | 1114 | ||
| AK122410.2 | - | 13 | 1109 | |||
| BB659027.1 | - | 1 | 348 | |||
| BC031745.1 | - | 13 | 1094 | 4506005 | ||
| BY713276.1 | - | 1 | 392 | |||
| X14836.1 | - | 13 | 1097 | |||
| X87142.1 | - | 1 | 392 | |||