Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000210.3 | NP_000201.2 | 4 | 655 | ||
NM_001079818.2 | NP_001073286.1 | 4 | 655 | |||
NM_001316306.1 | NP_001303235.1 | 4 | 356 | |||
XM_006712510.1 | XP_006712573.1 | 4 | 624 | |||
XM_006712511.1 | XP_006712574.1 | 4 | 624 | |||
XM_017004005.1 | XP_016859494.1 | 4 | 351 | |||
XM_017004006.1 | XP_016859495.1 | 4 | 351 | |||
XM_017004007.1 | XP_016859496.1 | 4 | 351 | |||
XM_017004008.1 | XP_016859497.1 | 4 | 351 | |||
GenBank mRNA | AB208842.1 | - | 4 | 323 | ||
AK296496.1 | - | 4 | 292 | |||
AK311695.1 | - | 4 | 356 | |||
BC007002.1 | - | 4 | 621 | |||
BC136455.1 | - | 4 | 529 | 9020444 | ||
BC136456.1 | - | 4 | 529 | 9020445 | ||
X53586.1 | - | 4 | 567 | |||
X59512.1 | - | 4 | 403 |