Search
Search

CPB2| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278541.1 | NP_001265470.1 | 8 | 834 | ||
| NM_001872.4 | NP_001863.3 | 9 | 945 | |||
| XM_017020393.1 | XP_016875882.1 | 9 | 888 | |||
| GenBank mRNA | AB011969.1 | - | 8 | 786 | ||
| AK290829.1 | - | 9 | 893 | |||
| BC007057.1 | - | 9 | 902 | 3934520 | ||
| BT006936.1 | - | 9 | 878 | |||
| M75106.1 | - | 9 | 897 | |||