Search
Search

PLPP3| Gene Symbol: PLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003713.4 | NP_003704.3 | 6 | 1487 | ||
| GenBank mRNA | AB000889.1 | - | 6 | 925 | ||
| AF017786.1 | - | 7 | 1293 | |||
| AF043329.1 | - | 6 | 1212 | |||
| AF480883.1 | - | 7 | 1299 | |||
| AK123105.1 | - | 2 | 1905 | |||
| AK312439.1 | - | 6 | 1445 | |||
| BC009196.2 | - | 6 | 1290 | 3960223 | ||
| BT019589.1 | - | 6 | 919 | |||
| U79294.1 | - | 7 | 1292 | |||