Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KYAT1 SIRNA Assays ›
          Gene Symbol:
          KYAT1
          Gene Name:
          kynurenine aminotransferase 1
          Gene Aliases:
          CCBL1, GTK, KAT1, KATI
          Chromosome Location:
          Chr.9: 128832942 - 128882551 on Build GRCh38
          UniGene ID:
          Hs.495250
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111417

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KYAT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122671.1 NP_001116143.1 3, 4 378
          NM_001287390.1 NP_001274319.1 5, 6 775
          NM_004059.4 NP_004050.3 3, 4 384
          NR_109829.1 3, 4 384
          XM_006717311.3 XP_006717374.1 4, 5 570
          XM_011519165.2 XP_011517467.1 5, 6 746
          XM_011519167.2 XP_011517469.2 3, 4 350
          XM_011519168.1 XP_011517470.1 2, 3 209
          XM_011519169.1 XP_011517471.1 3, 4 478
          XM_011519170.2 XP_011517472.1 3, 4 846
          XM_017015260.1 XP_016870749.1 4, 5 530
          XM_017015261.1 XP_016870750.1 5, 6 752
          XM_017015262.1 XP_016870751.1 5, 6 1241
          XM_017015263.1 XP_016870752.1 5, 6 656
          XM_017015264.1 XP_016870753.1 4, 5 563
          XM_017015265.1 XP_016870754.1 3, 4 940
          XM_017015266.1 XP_016870755.1 4, 5 571
          XM_017015267.1 XP_016870756.1 3, 4 392
          XM_017015268.1 XP_016870757.1 4, 5 554
          GenBank mRNA AK291178.1 - 3, 4 372
          AK297995.1 - 5, 6 631
          AK314427.1 - 3, 4 254
          AU123740.1 - 3, 4 362
          BC021262.2 - 3, 4 345 4137942
          DA381449.1 - 3, 4 384
          X82224.1 - 2, 3 199

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          L-phenylalanine catabolic process
          tryptophan catabolic process
          cellular modified amino acid metabolic process
          cellular amino acid biosynthetic process
          kynurenine metabolic process
          L-kynurenine catabolic process
          protein binding
          transaminase activity
          kynurenine-oxoglutarate transaminase activity
          pyridoxal phosphate binding
          protein homodimerization activity
          L-phenylalanine:pyruvate aminotransferase activity
          glutamine-phenylpyruvate transaminase activity
          cysteine-S-conjugate beta-lyase activity
          L-glutamine:pyruvate aminotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0