Search
Search

ADAMTS16| Gene Symbol: ADAMTS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139056.3 | NP_620687.2 | 15, 16 | 2423 | ||
| NR_136935.1 | 14, 15 | 2292 | ||||
| GenBank mRNA | AB095949.1 | - | 12, 13 | 1678 | ||
| AJ315734.1 | - | 15, 16 | 2285 | |||
| AK127746.1 | - | 15, 16 | 2423 | |||