Search
Search

ADAM32| Gene Symbol: ADAM32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145004.6 | NP_659441.3 | 11 | 1283 | ||
| XM_006716298.2 | XP_006716361.1 | 11 | 1077 | |||
| XM_011544433.1 | XP_011542735.1 | 11 | 1077 | |||
| XM_017013183.1 | XP_016868672.1 | 10 | 1008 | |||
| XM_017013184.1 | XP_016868673.1 | 11 | 1077 | |||
| XM_017013185.1 | XP_016868674.1 | 10 | 1083 | |||
| XM_017013186.1 | XP_016868675.1 | 7 | 687 | |||
| XM_017013187.1 | XP_016868676.1 | 3 | 223 | |||
| XM_017013188.1 | XP_016868677.1 | 2 | 192 | |||
| GenBank mRNA | AK131038.1 | - | 3 | 252 | ||
| AK313864.1 | - | 11 | 1104 | |||
| AY358739.1 | - | 11 | 1163 | |||
| BC026085.2 | - | 11 | 1067 | 4838404 | ||
| BC026169.2 | - | 11 | 1094 | 4829390 | ||
| BC028702.2 | - | 11 | 1077 | 4829385 | ||
| BC030014.2 | - | 11 | 1070 | 4830659 | ||
| BC030698.2 | - | 11 | 1085 | 4822988 | ||
| BC034975.2 | - | 11 | 1078 | 4829806 | ||