Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WT1 SIRNA Assays ›
          Gene Symbol:
          WT1
          Gene Name:
          Wilms tumor 1
          Gene Aliases:
          AWT1, EWS-WT1, GUD, NPHS4, WAGR, WIT-2, WT33
          Chromosome Location:
          Chr.11: 32387775 - 32435535 on Build GRCh38
          UniGene ID:
          Hs.591980
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 116364

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000378.4 NP_000369.3 7 1458
          NM_001198551.1 NP_001185480.1 8 961
          NM_001198552.1 NP_001185481.1 7 910
          NM_024424.3 NP_077742.2 8 1509
          NM_024426.4 NP_077744.3 8 1509
          GenBank mRNA AB971668.1 - 8 1668
          AK093168.1 - 7 906
          AK291736.1 - 8 1509
          BC032861.2 - 8 956 4827849
          BC046461.1 - 8 1514
          M30393.1 - 7 812
          S74529.1 - 128
          S75264.1 - 2 281
          X51630.1 - 8 1495

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_144783, M55512, DQ537939, BG087452.

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculogenesis
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          kidney development
          regulation of organ formation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          germ cell development
          thorax and anterior abdomen determination
          heart development
          sex determination
          negative regulation of cell proliferation
          RNA splicing
          gonad development
          male gonad development
          tissue development
          negative regulation of cell growth
          adrenal gland development
          male genitalia development
          epithelial cell differentiation
          glomerulus development
          glomerular basement membrane development
          adrenal cortex formation
          camera-type eye development
          negative regulation of apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          mesenchymal to epithelial transition
          positive regulation of heart growth
          diaphragm development
          cardiac muscle cell fate commitment
          visceral serous pericardium development
          cellular response to cAMP
          cellular response to gonadotropin stimulus
          metanephric mesenchyme development
          glomerular visceral epithelial cell differentiation
          posterior mesonephric tubule development
          metanephric epithelium development
          metanephric S-shaped body morphogenesis
          negative regulation of metanephric glomerular mesangial cell proliferation
          positive regulation of male gonad development
          negative regulation of female gonad development
          positive regulation of metanephric ureteric bud development
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          protein binding
          zinc ion binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          C2H2 zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0