Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMBT1 SIRNA Assays ›
Gene Symbol:
DMBT1
Gene Name:
deleted in malignant brain tumors 1
Gene Aliases:
GP340, SAG, muclin
Chromosome Location:
Chr.10: 122560665 - 122643736 on Build GRCh38
UniGene ID:
Hs.279611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118217
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMBT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320644.1
NP_001307573.1
41, 42
5335
NM_004406.2
NP_004397.2
28, 29
3451
NM_007329.2
NP_015568.2
41, 42
5335
NM_017579.2
NP_060049.2
40, 41
5305
XM_006717660.3
XP_006717723.1
43, 44
5662
XM_006717665.3
XP_006717728.1
42, 43
5614
XM_011539388.2
XP_011537690.1
44, 45
5710
XM_011539389.2
XP_011537691.1
44, 45
5710
XM_011539390.2
XP_011537692.1
43, 44
5680
XM_011539391.2
XP_011537693.1
43, 44
5680
XM_011539392.2
XP_011537694.1
43, 44
5680
XM_011539393.2
XP_011537695.1
43, 44
5680
XM_011539394.2
XP_011537696.1
43, 44
5680
XM_011539395.2
XP_011537697.1
43, 44
5680
XM_011539396.2
XP_011537698.1
43, 44
5662
XM_011539398.2
XP_011537700.1
42, 43
5647
XM_011539399.2
XP_011537701.1
42, 43
5614
XM_011539400.2
XP_011537702.1
43, 44
5386
XM_011539401.2
XP_011537703.1
41, 42
5323
XM_011539402.2
XP_011537704.1
41, 42
5323
XM_011539403.2
XP_011537705.1
41, 42
5323
XM_011539405.2
XP_011537707.1
41, 42
5324
XM_011539407.2
XP_011537709.1
41, 42
5317
XM_011539408.2
XP_011537710.1
41, 42
5317
XM_011539409.2
XP_011537711.1
41, 42
5317
XM_011539410.2
XP_011537712.1
41, 42
5317
XM_011539411.2
XP_011537713.1
40, 41
5293
XM_011539413.2
XP_011537715.1
38, 39
4929
XM_011539414.2
XP_011537716.1
28, 29
3430
XM_011539415.2
XP_011537717.1
26, 27
3384
XM_017015798.1
XP_016871287.1
29, 30
3972
GenBank mRNA
AF159456.1
-
41, 42
5335
AJ000342.1
-
28, 29
3451
AJ243212.1
-
41, 42
5335
AJ243224.1
-
40, 41
5305
AJ297935.1
-
41, 42
5316
AK304128.1
-
11, 12
1185
AK304149.1
-
15, 16
1475
BC153299.1
-
25, 26
3069
40147087
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
pattern recognition receptor signaling pathway
receptor-mediated endocytosis
multicellular organism development
protein transport
epithelial cell differentiation
induction of bacterial agglutination
cellular protein metabolic process
innate immune response
defense response to virus
scavenger receptor activity
protein binding
signaling pattern recognition receptor activity
zymogen binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gh6bl:80/100.66.76.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0