Search
Search

KCTD14
NDUFC2-KCTD14| Gene Symbol: KCTD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282406.1 | NP_001269335.1 | 3 | 513 | ||
| NM_023930.3 | NP_076419.2 | 2 | 402 | |||
| GenBank mRNA | AK313892.1 | - | 3 | 513 | ||
| BC001062.2 | - | 2 | 368 | 2819500 | ||
| BC001929.1 | - | 2 | 368 | 2819500 | ||
| CB993622.1 | - | 2 | 449 | |||
| Gene Symbol: NDUFC2-KCTD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001203260.1 | NP_001190189.1 | 4 | 1167 | ||
| NM_001203261.1 | NP_001190190.1 | 3 | 1072 | |||
| NM_001203262.1 | NP_001190191.1 | 3 | 1023 | |||