Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 6 | 1232 | ||
NM_001282151.1 | NP_001269080.1 | 3 | 841 | |||
NM_016226.4 | NP_057310.1 | 4 | 1077 | |||
NM_057180.2 | NP_476528.1 | 5 | 1089 | |||
NR_104099.1 | 6 | 1129 | ||||
NR_104100.1 | 5 | 1117 | ||||
NR_104101.1 | 5 | 1167 | ||||
NR_104102.1 | 4 | 968 | ||||
XM_006719459.3 | XP_006719522.1 | 6 | 1138 | |||
XM_006719460.3 | XP_006719523.1 | 7 | 1175 | |||
XM_011538484.2 | XP_011536786.1 | 5 | 1123 | |||
XM_017019457.1 | XP_016874946.1 | 5 | 1148 | |||
XM_017019458.1 | XP_016874947.1 | 4 | 1268 | |||
XM_017019459.1 | XP_016874948.1 | 4 | 930 | |||
GenBank mRNA | AF168716.1 | - | 5 | 1047 | ||
AK000499.1 | - | 3 | 805 | |||
AK130510.1 | - | 1875 | ||||
AL832866.1 | - | 4 | 3070 |