Search
Search

Ptk6| Gene Symbol: Ptk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009184.2 | NP_033210.1 | 3 | 743 | ||
| GenBank mRNA | AK154717.1 | - | 481 | |||
| BC125342.1 | - | 3 | 714 | 40129857 | ||
| BC125344.1 | - | 3 | 714 | 40129859 | ||
| BC145042.1 | - | 3 | 683 | |||
| U16805.1 | - | 3 | 743 | |||