Search
Search

Hif1a| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010431.2 | NP_034561.2 | 9 | 1574 | ||
| GenBank mRNA | AF003695.1 | - | 9 | 1408 | ||
| AK034087.1 | - | 9 | 1540 | |||
| AK076395.1 | - | 7 | 862 | |||
| AK138944.1 | - | 9 | 1441 | |||
| AK150367.1 | - | 9 | 1441 | |||
| X95580.1 | - | 9 | 1240 | |||