Search
Search

Exosc4| Gene Symbol: Exosc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175399.4 | NP_780608.1 | 3 | 1709 | ||
| GenBank mRNA | AK005101.2 | - | 3 | 1637 | ||
| AK083308.1 | - | 3 | 1670 | |||
| AK089890.1 | - | 3 | 1710 | |||
| AK144062.1 | - | 3 | 1624 | |||
| AK167629.1 | - | 3 | 1672 | |||