Search
Search

SNAI1| Gene Symbol: SNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005985.3 | NP_005976.2 | 2 | 530 | ||
| GenBank mRNA | AF125377.1 | - | 2 | 507 | ||
| AF131208.1 | - | 2 | 446 | |||
| AK313228.1 | - | 2 | 526 | |||
| BC012910.1 | - | 2 | 511 | 4537122 | ||
| BI822180.1 | - | 2 | 516 | |||
| DA972913.1 | - | 2 | 530 | |||