Search
Search

FADS1| Gene Symbol: FADS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013402.4 | NP_037534.3 | 2 | 541 | ||
| XM_011545022.2 | XP_011543324.1 | 2 | 215 | |||
| GenBank mRNA | AF084558.1 | - | 2 | 320 | ||
| AF199596.1 | - | 2 | 242 | |||
| AF226273.1 | - | 2 | 242 | |||
| AK027427.1 | - | 2 | 251 | |||
| AK027522.1 | - | 2 | 341 | |||
| AK074754.1 | - | 2 | 480 | |||
| AK074819.1 | - | 2 | 475 | |||
| AK096275.1 | - | 2 | 474 | |||
| AK222906.1 | - | 2 | 285 | |||
| AK289552.1 | - | 2 | 244 | |||
| AK298871.1 | - | 2 | 360 | |||
| AK314199.1 | - | 2 | 325 | |||
| AL512760.1 | - | 2 | 322 | |||
| BC007846.2 | - | 2 | 264 | 4129208 | ||
| BP253788.1 | - | 2 | 567 | |||