Search
Search

C21orf2| Gene Symbol: C21orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271440.1 | NP_001258369.1 | 1 | 222 | ||
| NM_001271441.1 | NP_001258370.1 | 1 | 222 | |||
| NM_004928.2 | NP_004919.1 | 1 | 222 | |||
| XM_006724053.3 | XP_006724116.1 | 1 | 75 | |||
| XM_017028470.1 | XP_016883959.1 | 1 | 237 | |||
| XM_017028471.1 | XP_016883960.1 | 1 | 103 | |||
| XM_017028472.1 | XP_016883961.1 | 1 | 157 | |||
| GenBank mRNA | BC031300.1 | - | 1 | 213 | 4878257 | |
| BC072012.1 | - | 1 | 195 | 6645626 | ||
| BM761336.1 | - | 1 | 222 | |||
| U84569.1 | - | 1 | 194 | |||
| Y11392.1 | - | 1 | 82 | |||
| Z93322.1 | - | 1 | 21 | |||