Search
Search

Pcgf1
LOC103692167| Gene Symbol: Pcgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007000.1 | NP_001007001.1 | 1 | 26 | ||
| GenBank mRNA | AY662670.1 | - | 1 | 26 | ||
| Gene Symbol: LOC103692167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763082.1 | XP_008761304.1 | 1 | 203 | ||